Learn More
Bioworld n-Dodecyl-β-D-Maltopyranoside, 5 g
Supplier:  Bioworld 404300172
Encompass_Preferred
Non-ionic detergent for solubilization of membrane proteins.Specification:Purity: ≥99.0%Absorbance (1%): <0.02 (340nm); <0.04 (280nm); <0.06 (260nm); <0.10 (225nm)n-dodecanol: ≤0.005%Conductance: <100μS (10% H2O)alpha isomer (NMR): <2%PH Value: 5.0 - 8.0 @ 25ºC ±0.1
Catalog No. 50-255-880