Learn More
Alkali Scientific n Dodecyl β D maltoside, 5 G
SDP
Catalog No. 5000661358
Encompass_Preferred
n-Dodecyl Maltoside is a nonionic detergent used for the gentle solubilization of membrane proteins. The 5g format is ideal for smaller-scale applications or as a starter quantity for solubilization tests. It is highly effective for isolating membrane-bound proteins without altering their functional integrity, making it ideal for proteomics, structural biology, and other protein analysis techniques. n-Dodecyl Maltoside disrupts lipid bilayers, allowing proteins to be extracted in their native state. It is often used in laboratories studying membrane proteins, protein-protein interactions, and protein structure-function relationships.
Catalog No. 50-006-61358 Supplier Alkali Scientific Supplier No. NDO004CP
n-Dodecyl Maltoside is a nonionic detergent used for the gentle solubilization of membrane proteins. The 5g format is ideal for smaller-scale applications or as a starter quantity for solubilization tests. It is highly effective for isolating membrane-bound proteins without altering their functional integrity, making it ideal for proteomics, structural biology, and other protein analysis techniques. n-Dodecyl Maltoside disrupts lipid bilayers, allowing proteins to be extracted in their native state. It is often used in laboratories studying membrane proteins, protein-protein interactions, and protein structure-function relationships.