Learn More
Medchemexpress LLC N-Dodecyl-β-D-maltoside | 69227-93-6 | 1 ML
SDP
Catalog No. 5000339513
Encompass_Preferred
N-Dodecyl-β-D-maltoside (Lauryl Maltoside) is a non-ionic detergent. It is known to promote the reactivation of various proteins and can effectively stabilize photoactive reaction center complexes (RCs), inhibiting their degradation.
  • Exhibits strong adsorption on alumina, titanium dioxide, and hematite.
  • Used for purification and stabilization of RNA polymerase.
  • Utilized for the detection of protein-lipid interactions.
  • Promotes reactivation of denatured adenosine deaminase, 3-phosphoglycerol phosphokinase, myokinase, and 3-hydroxysteroid dehydrogenase.
  • Effectively stabilizes photoactive reaction center complexes (RCs) in solution.
Catalog No. 50-003-39513 Supplier Medchemexpress LLC Supplier No. HY12897410MM/1ML
N-Dodecyl-β-D-maltoside (Lauryl Maltoside) is a non-ionic detergent. It is known to promote the reactivation of various proteins and can effectively stabilize photoactive reaction center complexes (RCs), inhibiting their degradation.
  • Exhibits strong adsorption on alumina, titanium dioxide, and hematite.
  • Used for purification and stabilization of RNA polymerase.
  • Utilized for the detection of protein-lipid interactions.
  • Promotes reactivation of denatured adenosine deaminase, 3-phosphoglycerol phosphokinase, myokinase, and 3-hydroxysteroid dehydrogenase.
  • Effectively stabilizes photoactive reaction center complexes (RCs) in solution.