Learn More
Medchemexpress LLC N-Dodecyl-β-D-maltoside | 69227-93-6 | 99.9% | 1 G
SDP
Catalog No. 5000338933
Encompass_Preferred
N-Dodecyl-β-D-maltoside (Lauryl Maltoside) is a non-ionic detergent. It exhibits strong adsorption on alumina, titanium dioxide, and hematite. This compound can promote the reactivation of various proteins and effectively stabilize photoactive reaction center complexes (RCs), inhibiting the degradation of Rhodopseudomonas spheroides R-26 reaction center in solution. It is suitable for the purification and stabilization of RNA polymerase and for detecting protein-lipid interactions.
  • Exhibits strong adsorption on alumina, titanium dioxide, and hematite.
  • Promotes reactivation of various proteins and stabilizes photoactive reaction center complexes.
  • Inhibits degradation of Rhodopseudomonas spheroides R-26 reaction center.
  • Suitable for purification and stabilization of RNA polymerase and detection of protein-lipid interactions.
  • Promotes reactivation of denatured enzymes, such as adenosine deaminase.
Catalog No. 50-003-38933 Supplier Medchemexpress LLC Supplier No. HY1289741G
N-Dodecyl-β-D-maltoside (Lauryl Maltoside) is a non-ionic detergent. It exhibits strong adsorption on alumina, titanium dioxide, and hematite. This compound can promote the reactivation of various proteins and effectively stabilize photoactive reaction center complexes (RCs), inhibiting the degradation of Rhodopseudomonas spheroides R-26 reaction center in solution. It is suitable for the purification and stabilization of RNA polymerase and for detecting protein-lipid interactions.
  • Exhibits strong adsorption on alumina, titanium dioxide, and hematite.
  • Promotes reactivation of various proteins and stabilizes photoactive reaction center complexes.
  • Inhibits degradation of Rhodopseudomonas spheroides R-26 reaction center.
  • Suitable for purification and stabilization of RNA polymerase and detection of protein-lipid interactions.
  • Promotes reactivation of denatured enzymes, such as adenosine deaminase.