Learn More
Medchemexpress LLC N-Dodecyl-β-D-maltoside | 69227-93-6 | 99.9% | 100 MG
SDP
Catalog No. 5000338794
Encompass_Preferred
N-Dodecyl-β-D-maltoside, also known as Lauryl Maltoside, is a non-ionic detergent. It is highly valued in biochemical research for its ability to promote protein reactivation and stabilize sensitive biological complexes.
  • Exhibits strong adsorption on alumina, titanium dioxide, and hematite.
  • Promotes the reactivation of various proteins.
  • Effectively stabilizes photoactive reaction center complexes (RCs).
  • Inhibits the degradation of Rhodopseudomonas spheroides R-26 reaction center in solution.
  • Suitable for the purification and stabilization of RNA polymerase.
  • Useful for detecting protein-lipid interactions.
Catalog No. 50-003-38794 Supplier Medchemexpress LLC Supplier No. HY128974100MG
N-Dodecyl-β-D-maltoside, also known as Lauryl Maltoside, is a non-ionic detergent. It is highly valued in biochemical research for its ability to promote protein reactivation and stabilize sensitive biological complexes.
  • Exhibits strong adsorption on alumina, titanium dioxide, and hematite.
  • Promotes the reactivation of various proteins.
  • Effectively stabilizes photoactive reaction center complexes (RCs).
  • Inhibits the degradation of Rhodopseudomonas spheroides R-26 reaction center in solution.
  • Suitable for the purification and stabilization of RNA polymerase.
  • Useful for detecting protein-lipid interactions.