Learn More
Cayman Chemical n-Dodecyl-β-D-maltoside, 69227-93-6, 5 g
SDP
Supplier:  Cayman Chemical 164945
Encompass_Preferred
A non-ionic detergent commonly used to solubilize membrane-associated proteins; helps retain the native conformation and activity of membrane-associated proteins, and facilitates the reforming of these proteins after denaturation
Catalog No. NC1233679