Learn More
Medchemexpress LLC N-Dodecyl-β-D-maltoside-d25 | 849110-74-3 | 98.7% | 10 MG
SDP
Catalog No. 5000338934
Encompass_Preferred
N-Dodecyl-β-D-maltoside-d25 (Lauryl Maltoside-d25) is a deuterium-labeled non-ionic detergent. It exhibits strong adsorption properties on alumina, titanium dioxide, and hematite. This compound effectively promotes protein reactivation, stabilizes photoactive reaction center complexes (RCs), and inhibits degradation of Rhodopseudomonas spheroides R-26 reaction center. It is also used for RNA polymerase purification and stabilization, and for detecting protein-lipid interactions.
  • Deuterium labeled compound.
  • Non-ionic detergent with strong adsorption properties.
  • Promotes protein reactivation.
  • Stabilizes photoactive reaction center complexes (RCs).
  • Inhibits degradation of Rhodopseudomonas spheroides R-26 reaction center.
  • Used for RNA polymerase purification and stabilization.
  • Used for detection of protein-lipid interactions.
  • Functions as a tracer or internal standard for quantitative analysis (NMR, GC-MS, LC-MS).
Catalog No. 50-003-38934 Supplier Medchemexpress LLC Supplier No. HY128974S10MG
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
N-Dodecyl-β-D-maltoside-d25 (Lauryl Maltoside-d25) is a deuterium-labeled non-ionic detergent. It exhibits strong adsorption properties on alumina, titanium dioxide, and hematite. This compound effectively promotes protein reactivation, stabilizes photoactive reaction center complexes (RCs), and inhibits degradation of Rhodopseudomonas spheroides R-26 reaction center. It is also used for RNA polymerase purification and stabilization, and for detecting protein-lipid interactions.
  • Deuterium labeled compound.
  • Non-ionic detergent with strong adsorption properties.
  • Promotes protein reactivation.
  • Stabilizes photoactive reaction center complexes (RCs).
  • Inhibits degradation of Rhodopseudomonas spheroides R-26 reaction center.
  • Used for RNA polymerase purification and stabilization.
  • Used for detection of protein-lipid interactions.
  • Functions as a tracer or internal standard for quantitative analysis (NMR, GC-MS, LC-MS).