Learn More
Medchemexpress LLC N-dodecyl-β-D-maltoside-d25 | 849110-74-3 | 98.7% | 535.77 g·mol⁻¹ | C24H21D25O11 | 5 MG
SDP
Catalog No. 5000263105
Encompass_Preferred
N-dodecyl-β-D-maltoside-d25 is a deuterium-labeled form of the non-ionic detergent DDM used to solubilize and stabilize membrane proteins and protein-lipid complexes in biochemical and biophysical studies. The deuterated isotopologue enables mass spectrometry and spectroscopic tracing while retaining gentle solubilizing properties.
  • Deuterium-labeled for mass spectrometry and spectroscopic applications.
  • Non-ionic detergent that preserves membrane protein activity.
  • High aqueous solubility (≈75 mg/mL); may require ultrasonic treatment.
  • Supplied as a powder with recommended cold storage for long-term stability.
  • Suitable for purification and stabilization of membrane complexes and RNA polymerase.
  • Facilitates identification of protein-lipid interactions and stabilization of photoactive complexes.
Catalog No. 50-002-63105 Supplier Medchemexpress LLC Supplier No. HY128974S5MG
N-dodecyl-β-D-maltoside-d25 is a deuterium-labeled form of the non-ionic detergent DDM used to solubilize and stabilize membrane proteins and protein-lipid complexes in biochemical and biophysical studies. The deuterated isotopologue enables mass spectrometry and spectroscopic tracing while retaining gentle solubilizing properties.
  • Deuterium-labeled for mass spectrometry and spectroscopic applications.
  • Non-ionic detergent that preserves membrane protein activity.
  • High aqueous solubility (≈75 mg/mL); may require ultrasonic treatment.
  • Supplied as a powder with recommended cold storage for long-term stability.
  • Suitable for purification and stabilization of membrane complexes and RNA polymerase.
  • Facilitates identification of protein-lipid interactions and stabilization of photoactive complexes.