Learn More
Medchemexpress LLC N-dodecyl-N,N-dimethyldodecan-1-aminium-d25 bromide | 918897-30-0 | 487.79 | 2.5 MG
SDP
Catalog No. 5000453204
Encompass_Preferred
N-Dodecyl-N,N-dimethyldodecan-1-aminium-d25 (bromide) is the deuterium labeled N-Dodecyl-N,N-dimethyldodecan-1-aminium bromide.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-53204 Supplier Medchemexpress LLC Supplier No. HYW008608S2.5MG
N-Dodecyl-N,N-dimethyldodecan-1-aminium-d25 (bromide) is the deuterium labeled N-Dodecyl-N,N-dimethyldodecan-1-aminium bromide.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for quantitation during the drug development process.
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.