Learn More
Cayman Chemical n-DodecylD-maltosIde 1g
SDP
Catalog No. 502969296
Encompass_Preferred
A non-ionic detergent commonly used to solubilize membrane-associated proteins; helps retain the native conformation and activity of membrane-associated proteins, and facilitates the reforming of these proteins after denaturation
Catalog No. 50-296-9296 Supplier Cayman Chemical Supplier No. 164941 G
A non-ionic detergent commonly used to solubilize membrane-associated proteins; helps retain the native conformation and activity of membrane-associated proteins, and facilitates the reforming of these proteins after denaturation