Learn More
Cayman Chemical n-Dodecyl-β-D-maltoside, Purity: ≥95%, CAS: 69227-93-6, 500 mg
SDP
Catalog No. 502970186
Encompass_Preferred
A non-ionic detergent commonly used to solubilize membrane-associated proteins; helps retain the native conformation and activity of membrane-associated proteins, and facilitates the reforming of these proteins after denaturation
Catalog No. 50-297-0186 Supplier Cayman Chemical Supplier No. 16494500 MG
A non-ionic detergent commonly used to solubilize membrane-associated proteins; helps retain the native conformation and activity of membrane-associated proteins, and facilitates the reforming of these proteins after denaturation