Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Dodecyltrimethoxysilane, 95%

Catalog No. AAL1648614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1648614 25 g
AAL1648622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1648614 Supplier Thermo Scientific Chemicals Supplier No. L1648614
  • $60.90
    $51.65 / Each of 1

    Save $9.25

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3069-21-4 | C15H34O3Si | 290.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3069-21-4
Molecular Formula C15H34O3Si
Molecular Weight (g/mol) 290.52
MDL Number MFCD00069148
InChI Key SCPWMSBAGXEGPW-UHFFFAOYSA-N
Synonym n-dodecyltrimethoxysilane, silane, dodecyltrimethoxy, lauryltrimethoxysilane, dodecyl trimethoxy silane, n-dodecytrimethoxysilane, acmc-1cr18, ksc494k8f
PubChem CID 76479
IUPAC Name dodecyl(trimethoxy)silane
SMILES CCCCCCCCCCCC[Si](OC)(OC)OC

Specifications

Boiling Point 125°C (1.8mmHg)
Melting Point -40°C
Quantity 25 g
Beilstein 2356323
Flash Point 108°C (226°F)
Formula Weight 290.52
Percent Purity 95%
Refractive Index 1.4274
Sensitivity Moisture sensitive
Density 0.89
Chemical Name or Material n-Dodecyltrimethoxysilane
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 221-332-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.