Learn More
Medchemexpress LLC N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]tetracosanamide | 34435-05-7 | 99.9% | C42H83NO3 | 10MG
SDP
Catalog No. 5000260017
Encompass_Preferred
C24-Ceramide is a long-chain sphingolipid (Cer(d18:1/24:0)) used in biochemical and membrane research. Supplied as a research-grade solid, it is applied in studies of membrane structure, lipid signaling, and lipid metabolism. Users should consult the supplier COA and SDS for batch-specific purity, solubility, and storage instructions.
Catalog No. 50-002-60017 Supplier Medchemexpress LLC Supplier No. HY13450810MG
C24-Ceramide is a long-chain sphingolipid (Cer(d18:1/24:0)) used in biochemical and membrane research. Supplied as a research-grade solid, it is applied in studies of membrane structure, lipid signaling, and lipid metabolism. Users should consult the supplier COA and SDS for batch-specific purity, solubility, and storage instructions.