Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]tetradecanamide | 34227-72-0 | 99.9% | 509.85 g/mol | C32H63NO3 | 5MG
SDP

Catalog No. 5000239045
Encompass_Preferred

C14 Ceramide is a high-purity sphingolipid (N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]tetradecanamide) used as a biochemical reagent for lipid research and cell signaling studies. The material is supplied as a powder for laboratory use and is provided with COA and SDS documentation for quality and safety verification.

  • High purity, 99.9%.
  • Molecular weight 509.85 g/mol.
  • Chemical formula C32H63NO3.
  • Suitable for biochemical assays and lipid analysis.
  • Supplied in small research pack sizes (mg scale).
  • Storage: powder at -20°C; in solution store at -80°C for long term.

Catalog No. 50-002-39045 Supplier Medchemexpress LLC Supplier No. HY1241775MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C14 Ceramide is a high-purity sphingolipid (N-[(E,2S,3R)-1,3-dihydroxyoctadec-4-en-2-yl]tetradecanamide) used as a biochemical reagent for lipid research and cell signaling studies. The material is supplied as a powder for laboratory use and is provided with COA and SDS documentation for quality and safety verification.

  • High purity, 99.9%.
  • Molecular weight 509.85 g/mol.
  • Chemical formula C32H63NO3.
  • Suitable for biochemical assays and lipid analysis.
  • Supplied in small research pack sizes (mg scale).
  • Storage: powder at -20°C; in solution store at -80°C for long term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.