Learn More
Medchemexpress LLC N-[(E)-[4-(diethylamino)-2-hydroxyphenyl]methylideneamino]pyridine-4-carboxamide | 140405-36-3 | MFCD00542031 | 99.7% | 312.37 g/mol | C17H20N4O2 | 50 MG
SDP
Catalog No. 5000245949
Encompass_Preferred
RSVA405 is a small-molecule research compound that potently activates AMP-activated protein kinase (AMPK) and is orally active in vivo. It facilitates CaMKKβ-dependent AMPK activation (EC50 = 1 μM), inhibits mTOR signaling, promotes autophagy and amyloid-β (Aβ) degradation, and shows anti-inflammatory activity via STAT3 inhibition.
  • Potent AMPK activator (EC50 = 1 μM).
  • Orally active small-molecule research compound.
  • Promotes autophagy and increases Aβ degradation.
  • Inhibits mTOR signaling and STAT3-mediated inflammation.
  • High purity (≈99.7%) light yellow solid.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-45949 Supplier Medchemexpress LLC Supplier No. HY10323850MG
RSVA405 is a small-molecule research compound that potently activates AMP-activated protein kinase (AMPK) and is orally active in vivo. It facilitates CaMKKβ-dependent AMPK activation (EC50 = 1 μM), inhibits mTOR signaling, promotes autophagy and amyloid-β (Aβ) degradation, and shows anti-inflammatory activity via STAT3 inhibition.
  • Potent AMPK activator (EC50 = 1 μM).
  • Orally active small-molecule research compound.
  • Promotes autophagy and increases Aβ degradation.
  • Inhibits mTOR signaling and STAT3-mediated inflammation.
  • High purity (≈99.7%) light yellow solid.
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).