Learn More
Medchemexpress LLC HY-128676 10mg , N-ε-propargyloxycarbonyl-L-lysine (hydrochloride) CAS:1428330-91-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY12867610MG
Encompass_Preferred
HY-128676 10mg N-ε-propargyloxycarbonyl-L-lysine (hydrochloride) CAS: 1428330-91-9 N-ε-propargyloxycarbonyl-L-lysine (H-L-Lys(Poc)-OH) hydrochloride is a lysine-based unnatural amino acid (UAA). N-ε-propargyloxycarbonyl-L-lysine is widely used for bio-conjugation of fluorescent probes in diverse organisms from E. coli to mammalian cells even in animals. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1736