Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-epsilon-Acetyl-L-lysine, 99%, MP Biomedicals™

Catalog No. ICN15023505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15023505 5 g
ICN15023501 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15023505 Supplier MP Biomedicals, Inc Supplier No. 0215023505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 692-04-6 | C8H16N2O3 | 188.23 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 692-04-6
Molecular Formula C8H16N2O3
Molecular Weight (g/mol) 188.23
MDL Number MFCD00002639
InChI Key DTERQYGMUDWYAZ-ZETCQYMHSA-N
Synonym n-epsilon-acetyl-l-lysine, n6-acetyl-l-lysine, nepsilon-acetyl-l-lysine, n-epsilon-acetyllysine, h-lys ac-oh, n6-acetyllysine, s-6-acetamido-2-aminohexanoic acid, n 6-acetyl-l-lysine, n 6-acetyllysine, n epsilon-acetyl-l-lysine
PubChem CID 92832
ChEBI CHEBI:17752
SMILES CC(=O)NCCCC[C@H](N)C(O)=O

Specifications

Color White
Assay Percent Range 99%
IUPAC Name (2S)-6-acetamido-2-aminohexanoic acid
Quantity 5 g
Formula Weight 188.3
Physical Form Powder
Chemical Name or Material N-epsilon-acetyl-L-lysine
Recommended Storage Store at 0°C.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.