Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-epsilon-FMOC-N-alpha-BOC-L-Lysine, 99%

Catalog No. AC439080050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC439080050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC439080050 Supplier Thermo Scientific Chemicals Supplier No. 439080050
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 84624-27-1 | C26H32N2O6 | 468.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84624-27-1
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.55
MDL Number MFCD00062032
InChI Key JYEVQYFWINBXJU-ANBDAQEENA-N
Synonym boc-lys fmoc-oh, n-boc-n'-fmoc-l-lysine, n2-boc-n6-fmoc-l-lysine, n-boc-n-fmoc-l-lysine, boclys fmoc, n-epsilon-fmoc-alpha-t-boc-l-lysine, 2s-2-tert-butoxycarbonyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, pubchem14939, n, a-boc-n, a-fmoc-l-lysine
PubChem CID 10983524
IUPAC Name (2S)-6-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 135°C to 138°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 5 g
Specific Rotation 3.4
Formula Weight 468.55
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-ε-FMOC-N-α-BOC-L-Lysine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.