Learn More
Medchemexpress LLC N-(ethoxycarbonylmethyl)-6-methoxyquinolinium bromide | 162558-52-3 | 99.8% | 200 MG
SDP
Catalog No. 5000410828
Encompass_Preferred
MQAE is a chloride ion (Cl-) fluorescent probe designed for measuring chloride concentrations. Its fluorescence intensity decreases proportionally as chloride ion levels increase, making it suitable for various detection methods like confocal microscopy and flow cytometry (Ex/Em=350/460 nm). It exhibits high cell permeability and is intended for research use only.
  • Suitable for measuring chloride concentrations
  • Fluorescence intensity decreases with increasing chloride ions
  • High cell permeability
  • Compatible with confocal microscopy and flow cytometry
Catalog No. 50-004-10828 Supplier Medchemexpress LLC Supplier No. HYD0090200MG
MQAE is a chloride ion (Cl-) fluorescent probe designed for measuring chloride concentrations. Its fluorescence intensity decreases proportionally as chloride ion levels increase, making it suitable for various detection methods like confocal microscopy and flow cytometry (Ex/Em=350/460 nm). It exhibits high cell permeability and is intended for research use only.
  • Suitable for measuring chloride concentrations
  • Fluorescence intensity decreases with increasing chloride ions
  • High cell permeability
  • Compatible with confocal microscopy and flow cytometry