Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-ethyl-1,6-dihydro-1,2-dimethyl-6-(methylimino)-N-phenyl-4-pyrimidinamine, monohydrochloride | 133059-99-1 | MFCD00914937 | 99.8% | 292.81 g/mol | C15H21ClN4 | 50 MG
SDP

Catalog No. 5000211639
Encompass_Preferred

ZD7288 (ICI D7288) is a selective hyperpolarization-activated cyclic nucleotide-gated (HCN) channel blocker used in electrophysiology and pharmacology research to inhibit HCN-mediated currents. It is supplied as a white to off-white solid with reported high purity and is intended for research use only.

  • Selective HCN channel blocker used for electrophysiology and pharmacology research.
  • High reported purity (≈99.8%) for consistent experimental results.
  • Molecular weight 292.81 g/mol for accurate dosing and calculations.
  • White to off-white solid appearance suitable for standard lab handling.
  • Recommended storage: sealed at 4°C away from moisture; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-11639 Supplier Medchemexpress LLC Supplier No. HY10134650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZD7288 (ICI D7288) is a selective hyperpolarization-activated cyclic nucleotide-gated (HCN) channel blocker used in electrophysiology and pharmacology research to inhibit HCN-mediated currents. It is supplied as a white to off-white solid with reported high purity and is intended for research use only.

  • Selective HCN channel blocker used for electrophysiology and pharmacology research.
  • High reported purity (≈99.8%) for consistent experimental results.
  • Molecular weight 292.81 g/mol for accurate dosing and calculations.
  • White to off-white solid appearance suitable for standard lab handling.
  • Recommended storage: sealed at 4°C away from moisture; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.