Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethyl-3-methylbenzamide, 97%

Catalog No. AAH5987103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5987103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5987103 Supplier Thermo Scientific Chemicals Supplier No. H5987103

missing translation for 'validMainframeMsg'

CAS: 26819-07-8 | C10H13NO | 163.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26819-07-8
Molecular Formula C10H13NO
Molecular Weight (g/mol) 163.22
MDL Number MFCD00496601
InChI Key FPINATACRXASTP-UHFFFAOYSA-N
Synonym benzamide, n-ethyl-3-methyl, n-ethyl-m-toluamide, benzamide,n-ethyl-3-methyl, benzamide, 3-methyl-n-ethyl, n-ethyl-3-methylbenzamide, benzamide, n-ethyl-3-methyl-9ci
PubChem CID 93103
IUPAC Name N-ethyl-3-methylbenzamide
SMILES CCNC(=O)C1=CC(=CC=C1)C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 163.22
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Ethyl-3-methylbenzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.