Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethyl-5-phenylisoxazolium-3'-sulfonate 95%, Thermo Scientific™

Catalog No. AC118840050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC118840050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC118840050 Supplier Thermo Scientific Chemicals Supplier No. 118840050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4156-16-5
Molecular Formula C11H11NO4S
Molecular Weight (g/mol) 253.272
MDL Number MFCD00012128
InChI Key MWOOKDULMBMMPN-UHFFFAOYSA-N
Synonym woodward's reagent k, n-ethyl-5-phenylisoxazolium-3'-sulfonate, 2-ethyl-5-phenylisoxazolium-3'-sulfonate, nepis, unii-3bn309r76h, 2-ethyl-5-3-sulfonatophenyl-1,2-oxazol-2-ium, woodward's reagent potassium, 2-ethyl-5-phenylisoxazolium-3'-sulphonate, n-ethyl-5-phenylisoxazolium-3/'-sulfonate
PubChem CID 77804
IUPAC Name 3-(2-ethyl-1,2-oxazol-2-ium-5-yl)benzenesulfonate
SMILES CC[N+]1=CC=C(O1)C2=CC(=CC=C2)S(=O)(=O)[O-]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Quantity 5 g
Fieser 01,384; 02,197; 05,306; 08,226
Merck Index 15, 10248
Formula Weight 253.27
Percent Purity ≥94%
Physical Form Crystalline Powder
Chemical Name or Material N-Ethyl-5-phenylisoxazolium-3'-sulfonate, 95%
Show More Show Less
EINECSNumber 223-988-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.