Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethyl-N-(4-hydroxybutyl)nitrosamine 98.0+%, TCI America™
SDP

Catalog No. E03075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03075G 5 g
E03071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03075G Supplier TCI America Supplier No. E03075G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 54897-62-0
Molecular Formula C6H14N2O2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00059606
InChI Key SAWPRHGGVOPPSR-UHFFFAOYSA-N
Synonym N-Ethyl-N-butan-4-ol-nitrosamine, N-Ethyl-N-nitroso-4-aminobutanol
PubChem CID 41223
IUPAC Name N-ethyl-N-(4-hydroxybutyl)nitrous amide
SMILES CCN(CCCCO)N=O

Specifications

Boiling Point 140°C
Quantity 5 g
Formula Weight 146.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Ethyl-N-(4-hydroxybutyl)nitrosamine
RTECSNumber EL3870000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.