Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethyl-N-methylcarbamoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. E096325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E096325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E096325G Supplier TCI America Supplier No. E096325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42252-34-6
Molecular Formula C4H8ClNO
Molecular Weight (g/mol) 121.56
MDL Number MFCD08061404
InChI Key XZVYDRLPXWFRIS-UHFFFAOYSA-N
PubChem CID 11240471
IUPAC Name N-ethyl-N-methylcarbamoyl chloride
SMILES CCN(C)C(Cl)=O

Specifications

Boiling Point 89°C
Color Yellow
UN Number 3265
Quantity 25 g
Formula Weight 121.56
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Ethyl-N-methylcarbamoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.