Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N'-Ethyl-N,N-dimethylethylenediamine, 98%

Catalog No. AAL0318006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0318006 5 g
AAL0318014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0318006 Supplier Thermo Scientific Chemicals Supplier No. L0318006
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 123-83-1 | C6H18N2 | 118.22 g/mol

Reatant in the synthesis of tripod amphiphile. N'-Ethyl-N,N-dimethylethylenediamine is used as catalysts for olefin aziridination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of tripod amphiphile. N′-Ethyl-N,N-dimethylethylenediamine is used as catalysts for olefin aziridination.

Notes
Air Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, carbondioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-83-1
Molecular Formula C6H18N2
Molecular Weight (g/mol) 118.22
MDL Number MFCD00009034
InChI Key WLNSKTSWPYTNLY-UHFFFAOYSA-P
Synonym n,n-dimethyl-n'-ethylethylenediamine, 1,2-ethanediamine, n'-ethyl-n,n-dimethyl, 2-ethylaminoethyldimethylamine, 2-dimethylamino ethyl ethyl amine, ethylenediamine, n'-ethyl-n,n-dimethyl, n'-ethyl-n,n-dimethylethylenediamine, n1-ethyl-n2,n2-dimethylethane-1,2-diamine, 1,2-ethanediamine, n2-ethyl-n1,n1-dimethyl, n1-ethyl-n2,n2-dimethyl-1,2-ethanediamine, n'-ethyl-n,n-dimethylethane-1,2-diamine
PubChem CID 67162
IUPAC Name N-ethyl-N',N'-dimethylethane-1,2-diamine
SMILES CC[NH2+]CC[NH+](C)C

Specifications

Density 0.818
Boiling Point 134°C to 135°C
Flash Point 23°C (73°F)
Odor Amine-like
Refractive Index 1.4225
Quantity 5 g
UN Number UN2734
Beilstein 773675
Formula Weight 116.21
Percent Purity 98%
Chemical Name or Material N'-Ethyl-N,N-dimethylethylenediamine
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 204-656-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.