Learn More
Medchemexpress LLC N-Ethyl-N N-dipropyl 5g
SDP
Catalog No. 5000206453
Encompass_Preferred
N-Ethyl-N N-dipropylpropan-1-aminium iodide is a quaternary ammonium compound belonging to the class of alkylammonium salts N-Ethyl-N N-dipropylpropan-1-aminium (iodide) is commonly used as a phase transfer catalyst in organic synthesis to transfer reactants between immiscible phases It can also be used as a surfactant antimicrobial or corrosion inhibitor Its unique chemical properties make it an essential reagent in many different industrial applications including pharmaceuticals agrochemicals and materials science
Catalog No. 50-002-06453 Supplier Medchemexpress LLC Supplier No. HYW0999295G
N-Ethyl-N N-dipropylpropan-1-aminium iodide is a quaternary ammonium compound belonging to the class of alkylammonium salts N-Ethyl-N N-dipropylpropan-1-aminium (iodide) is commonly used as a phase transfer catalyst in organic synthesis to transfer reactants between immiscible phases It can also be used as a surfactant antimicrobial or corrosion inhibitor Its unique chemical properties make it an essential reagent in many different industrial applications including pharmaceuticals agrochemicals and materials science