Learn More
Medchemexpress LLC N-Ethyl-N-nitrosoure | 759-73-9 | 10g
SDP
Catalog No. 5000301502
Encompass_Preferred
N-Ethyl-N-nitrosourea (ENU) is a DNA alkylating agent N-Ethyl-N-nitrosourea induces leukemia by alkylating nucleobases disrupting DNA and resulting in bone marrow suppression and the formation of leukemic cells N-Ethyl-N-nitrosourea is teratogenic in vivo inducing tumor formation and paw malformations in pregnant rats N-Ethyl-N-nitrosourea cause central nervous system (CNS) tumors and genetic disorders[1][2]
Catalog No. 50-003-01502 Supplier Medchemexpress LLC Supplier No. HYW01981510G
N-Ethyl-N-nitrosourea (ENU) is a DNA alkylating agent N-Ethyl-N-nitrosourea induces leukemia by alkylating nucleobases disrupting DNA and resulting in bone marrow suppression and the formation of leukemic cells N-Ethyl-N-nitrosourea is teratogenic in vivo inducing tumor formation and paw malformations in pregnant rats N-Ethyl-N-nitrosourea cause central nervous system (CNS) tumors and genetic disorders[1][2]