Learn More
Medchemexpress LLC N-ethyl-N-nitrosourea | 759-73-9 | MFCD00053635 | 99.9% | 117.11 | C3H7N3O2 | 5 G
SDP
Catalog No. 5000301500
Encompass_Preferred
N-ethyl-N-nitrosourea (ENU) is a potent DNA-alkylating reagent used in research to induce point mutations and study mutagenesis, carcinogenesis, and developmental toxicity. Supplied as a high-purity solid, ENU requires controlled storage and handling because of its teratogenic and carcinogenic properties.
  • High purity (99.9%) suitable for research applications.
  • Effective alkylating agent for inducing point mutations in genetic studies.
  • Available in multiple pack sizes, including 5 g, for laboratory use.
  • Solid, off-white to pink appearance for easy weighing and handling under precautions.
  • Storage under nitrogen at -20°C recommended; in solvent, -80°C for extended storage.
  • Includes safety and handling guidance due to teratogenic and carcinogenic effects.
Catalog No. 50-003-01500 Supplier Medchemexpress LLC Supplier No. HYW0198155G
N-ethyl-N-nitrosourea (ENU) is a potent DNA-alkylating reagent used in research to induce point mutations and study mutagenesis, carcinogenesis, and developmental toxicity. Supplied as a high-purity solid, ENU requires controlled storage and handling because of its teratogenic and carcinogenic properties.
  • High purity (99.9%) suitable for research applications.
  • Effective alkylating agent for inducing point mutations in genetic studies.
  • Available in multiple pack sizes, including 5 g, for laboratory use.
  • Solid, off-white to pink appearance for easy weighing and handling under precautions.
  • Storage under nitrogen at -20°C recommended; in solvent, -80°C for extended storage.
  • Includes safety and handling guidance due to teratogenic and carcinogenic effects.