Learn More
Medchemexpress LLC N-ethyl-N-oxido-dodecan-1-amine | 1643-20-5 | MFCD00002049 | 98.0% | 229.40 g/mol | C14H31NO | 5 G
SDP
Catalog No. 5000283092
Encompass_Preferred
N-ethyl-N-oxido-dodecan-1-amine is an amine oxide surfactant supplied as a research-grade reagent for laboratory and formulation use. It functions as a detergent, emulsifier, and wetting agent and is suitable for formulation studies, analytical work, and life-science research. Typical specifications include 98.0% purity, molecular formula C14H31NO, molecular weight 229.40 g/mol, and CAS 1643-20-5; available in a 5 g pack.
  • High-purity reagent for research and analytical applications.
  • Acts as a detergent, emulsifier, and wetting agent.
  • Suitable for formulation and laboratory studies.
  • Documented specification available: SDS, COA, data sheet, and H NMR.
  • Packaged for laboratory use in a 5 G vial.
Catalog No. 50-002-83092 Supplier Medchemexpress LLC Supplier No. HYW1277855G
N-ethyl-N-oxido-dodecan-1-amine is an amine oxide surfactant supplied as a research-grade reagent for laboratory and formulation use. It functions as a detergent, emulsifier, and wetting agent and is suitable for formulation studies, analytical work, and life-science research. Typical specifications include 98.0% purity, molecular formula C14H31NO, molecular weight 229.40 g/mol, and CAS 1643-20-5; available in a 5 g pack.
  • High-purity reagent for research and analytical applications.
  • Acts as a detergent, emulsifier, and wetting agent.
  • Suitable for formulation and laboratory studies.
  • Documented specification available: SDS, COA, data sheet, and H NMR.
  • Packaged for laboratory use in a 5 G vial.