Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethyl-o-toluidine, 98%, Thermo Scientific™

Catalog No. AAA11315AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA11315AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA11315AE Supplier Thermo Scientific Chemicals Supplier No. A11315AE

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-68-8
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00026695
InChI Key MWOUGPLLVVEUMM-UHFFFAOYSA-N
Synonym n-ethyl-o-toluidine, 2-ethylamino toluene, 2-ethylaminotoluene, benzenamine, n-ethyl-2-methyl, o-methyl-n-ethylaniline, 1-ethylamino-2-methylbenzene, n-ethyl-2-methyl-aniline, n-ethyl-2-aminotoluene, o-toluidine, n-ethyl, n-ethyl-2-toluidine
PubChem CID 7201
IUPAC Name N-ethyl-2-methylaniline
SMILES CCNC1=CC=CC=C1C

Specifications

Density 0.938
Boiling Point 218°C
Flash Point 98°C (208°F)
Refractive Index 1.546
Quantity 100 mL
UN Number UN2754
Beilstein 2689722
Formula Weight 135.21
Percent Purity 98%
Chemical Name or Material N-Ethyl-o-toluidine
Hazard Category H227-H301+H311-H330-H373
Hazard Statement GHS H Statement
H300-H310-H330-H373-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: N-ETHYLTOLUIDINES
EINECSNumber 202-354-3
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.