Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Ethyldiisopropylamine

Catalog No. 03440250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-440-250ML 250 mL
03-440-50ML 50 mL
03-440-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 03-440-250ML Supplier Sigma Aldrich Supplier No. 03440250ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N,N-Diisopropylethylamine (DIPEA), also known as Hunig'sas base, is a sterically hindered amine. It is a non-nucleophilic base commonly employed in substitution reactions, alkylations, and amide couplings, etc. DIPEA is also used as a base in the Pd catalyzed cross-coupling reactions, which include Heck coupling and Sonagashira coupling reactions.

Specifications

CAS 7087-68-5
Density 0.755 g/mL at 20°C, 0.755 g/mL at 20°C, 0.757 g/mL at 25°C
Boiling Point 126°C to 128°C
Molecular Formula C8H19N
Refractive Index n20/D 1.414
Linear Formula CH3CH2N[CH(CH3)2]2
MDL Number MFCD00008868
Quantity 250 mL
Synonym N,N-Diisopropylethylamine; H++nigs base
Molecular Weight (g/mol) 129.24 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.