Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-ethylglycine | 627-01-0 | MFCD00037794 | 98.0% | 103.12 g/mol | C4H9NO2 | 25 G
SDP

Catalog No. 5000282362
Encompass_Preferred

N-ethylglycine is an N-alkylated glycine derivative used as a reagent in biochemical and analytical research. Supplied as a solid, it is suitable for synthesis, assay development, and analytical workflows, with supporting documentation available.

  • High purity (98.0%).
  • Solid powder, packaged as a 25 g pack.
  • CAS number 627-01-0.
  • Molecular formula C4H9NO2; molecular weight 103.12 g/mol.
  • Suitable for synthesis, assay development, and analytical applications.
  • Datasheet and safety data sheet available.

Catalog No. 50-002-82362 Supplier Medchemexpress LLC Supplier No. HYW14187925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-ethylglycine is an N-alkylated glycine derivative used as a reagent in biochemical and analytical research. Supplied as a solid, it is suitable for synthesis, assay development, and analytical workflows, with supporting documentation available.

  • High purity (98.0%).
  • Solid powder, packaged as a 25 g pack.
  • CAS number 627-01-0.
  • Molecular formula C4H9NO2; molecular weight 103.12 g/mol.
  • Suitable for synthesis, assay development, and analytical applications.
  • Datasheet and safety data sheet available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.