Learn More
Medchemexpress LLC N-Ethylmaleimide | 128-53-0 | 200 MG
SDP
Catalog No. 5000410633
Encompass_Preferred
N-Ethylmaleimide (NEM) is derived from maleic acid and can alkylate free sulfhydryl groups. It acts as an irreversible cysteine protease inhibitor. NEM specifically inhibits phosphate transport in mitochondria and inhibits prolyl endopeptidase with an IC50 value of 6.3 μM. It can be used to modify cysteine residues in proteins and peptides, and is also a deubiquitinating enzyme inhibitor.
  • Irreversible cysteine protease inhibitor
  • Specifically inhibits phosphate transport in mitochondria
  • Inhibits prolyl endopeptidase (IC50 value of 6.3 μM)
  • Modifies cysteine residues in proteins and peptides
  • Deubiquitinating enzyme inhibitor
  • Inhibits Akt phosphorylation
  • Affects pro-caspase-3 conversion
  • Promotes vascular smooth muscle cell apoptosis
  • Affects PP2A activity and ROS production
Catalog No. 50-004-10633 Supplier Medchemexpress LLC Supplier No. HYD0843200MG
N-Ethylmaleimide (NEM) is derived from maleic acid and can alkylate free sulfhydryl groups. It acts as an irreversible cysteine protease inhibitor. NEM specifically inhibits phosphate transport in mitochondria and inhibits prolyl endopeptidase with an IC50 value of 6.3 μM. It can be used to modify cysteine residues in proteins and peptides, and is also a deubiquitinating enzyme inhibitor.
  • Irreversible cysteine protease inhibitor
  • Specifically inhibits phosphate transport in mitochondria
  • Inhibits prolyl endopeptidase (IC50 value of 6.3 μM)
  • Modifies cysteine residues in proteins and peptides
  • Deubiquitinating enzyme inhibitor
  • Inhibits Akt phosphorylation
  • Affects pro-caspase-3 conversion
  • Promotes vascular smooth muscle cell apoptosis
  • Affects PP2A activity and ROS production