Learn More
Medchemexpress LLC N-Ethylmaleimide | 128-53-0 | 50 MG
SDP
Catalog No. 5000410004
Encompass_Preferred
N-Ethylmaleimide (NEM) is a maleic acid derivative that functions by alkylating free sulfhydryl groups. It is recognized as an irreversible cysteine protease inhibitor and plays a role in inhibiting phosphate transport within mitochondria. The compound also inhibits prolyl endopeptidase with an IC50 value of 6.3 μM. It can be used to modify cysteine residues in proteins and peptides.
  • Alkylated free sulfhydryl groups
  • Irreversible cysteine protease inhibitor
  • Specifically inhibits phosphate transport in mitochondria
  • Inhibits prolyl endopeptidase with an IC50 value of 6.3 μM
  • Can be used to modify cysteine residues in proteins and peptides
Catalog No. 50-004-10004 Supplier Medchemexpress LLC Supplier No. HYD084350MG
N-Ethylmaleimide (NEM) is a maleic acid derivative that functions by alkylating free sulfhydryl groups. It is recognized as an irreversible cysteine protease inhibitor and plays a role in inhibiting phosphate transport within mitochondria. The compound also inhibits prolyl endopeptidase with an IC50 value of 6.3 μM. It can be used to modify cysteine residues in proteins and peptides.
  • Alkylated free sulfhydryl groups
  • Irreversible cysteine protease inhibitor
  • Specifically inhibits phosphate transport in mitochondria
  • Inhibits prolyl endopeptidase with an IC50 value of 6.3 μM
  • Can be used to modify cysteine residues in proteins and peptides