Learn More
Cayman Chemical N-EthylmaleImIde 5g
SDP
Catalog No. 502983402
Encompass_Preferred
A modifier of protein sulfhydryl groups; has commonly been used to inactivate cysteine-containing enzymes in vitro; inhibits prolyl endopeptidase (IC50 = 6.3 µM); alkylates Cys519 in Kv7.4 and activates Kv7.2, Kv7.4, and Kv7.5 in CHO cells at 50 µM
Catalog No. 50-298-3402 Supplier Cayman Chemical Supplier No. 199385 G
A modifier of protein sulfhydryl groups; has commonly been used to inactivate cysteine-containing enzymes in vitro; inhibits prolyl endopeptidase (IC50 = 6.3 µM); alkylates Cys519 in Kv7.4 and activates Kv7.2, Kv7.4, and Kv7.5 in CHO cells at 50 µM