Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Ethylsuccinimide 98.0+%, TCI America™
SDP

Catalog No. E017525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E017525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E017525G Supplier TCI America Supplier No. E017525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2314-78-5
Molecular Formula C6H9NO2
Molecular Weight (g/mol) 127.143
MDL Number MFCD00059748
InChI Key GHAZCVNUKKZTLG-UHFFFAOYSA-N
PubChem CID 16842
ChEBI CHEBI:44348
IUPAC Name 1-ethylpyrrolidine-2,5-dione
SMILES CCN1C(=O)CCC1=O

Specifications

Color White
Quantity 25 g
Formula Weight 127.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material N-Ethylsuccinimide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.