Learn More
Medchemexpress LLC HY-N2232 1mg Medchemexpress, N-​Feruloyloctopamine CAS:66648-44-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN22321MG
Encompass_Preferred
Medchemexpress, HY-N2232 1mg N-​Feruloyloctopamine CAS:66648-44-0 N-Feruloyloctopamine, isolated from Garlic skin, is an antioxidant constituent. N-Feruloyloctopamine significantly decreases the phosphorylation levels of Akt and p38 MAPK. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2879