Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-(fluoromethyl)-2-hydroxy-N,N-dimethylethanaminium chloride | 459424-38-5 | MFCD07995053 | 98.0% | 157.61 | C5H13ClFNO | 5 G
SDP

Catalog No. 5000241138
Encompass_Preferred

Fluorocholine chloride is a biochemical reagent (CAS 459424-38-5) used in life-science research and as a building block in biochemical and radiochemical studies. It is supplied as a solid or as solutions in DMSO and is characterized by a defined molecular composition and high purity.

  • Used as a biochemical reagent for life-science research.
  • Available as solid and DMSO solutions.
  • Molecular formula C5H13ClFNO; molecular weight 157.61.
  • Purity approximately 98.0%.
  • White to off-white solid appearance.
  • Store solids at 4°C; store solutions at -80°C for long-term.
  • Pack sizes available from milligram to multi-gram quantities.

Catalog No. 50-002-41138 Supplier Medchemexpress LLC Supplier No. HY1073515G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorocholine chloride is a biochemical reagent (CAS 459424-38-5) used in life-science research and as a building block in biochemical and radiochemical studies. It is supplied as a solid or as solutions in DMSO and is characterized by a defined molecular composition and high purity.

  • Used as a biochemical reagent for life-science research.
  • Available as solid and DMSO solutions.
  • Molecular formula C5H13ClFNO; molecular weight 157.61.
  • Purity approximately 98.0%.
  • White to off-white solid appearance.
  • Store solids at 4°C; store solutions at -80°C for long-term.
  • Pack sizes available from milligram to multi-gram quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.