Learn More
Medchemexpress LLC N-Fmoc-1,6-diaminohexane hydrochloride | 945923-91-1 | MFCD00830743 | 99.8% | 374.90 g·mol⁻¹ | C21H27ClN2O2 | 250 MG
SDP
Catalog No. 5000244697
Encompass_Preferred
Fmoc-1,6-diaminohexane hydrochloride is an Fmoc-protected primary diamine hydrochloride salt used as a building block and linker in solid-phase peptide synthesis and small-molecule conjugation. It is provided as a crystalline solid and is characterized by CAS 945923-91-1, molecular formula C21H27ClN2O2, and molecular weight 374.90 g·mol⁻¹.
  • Fmoc-protected primary diamine for selective deprotection.
  • Hydrochloride salt form improves stability and handling.
  • High reported purity for synthesis and analytical use.
  • Available in small lab-scale pack sizes for research use.
  • Characterized by CAS number and supported by certificate of analysis.
Catalog No. 50-002-44697 Supplier Medchemexpress LLC Supplier No. HY103664A250MG
Fmoc-1,6-diaminohexane hydrochloride is an Fmoc-protected primary diamine hydrochloride salt used as a building block and linker in solid-phase peptide synthesis and small-molecule conjugation. It is provided as a crystalline solid and is characterized by CAS 945923-91-1, molecular formula C21H27ClN2O2, and molecular weight 374.90 g·mol⁻¹.
  • Fmoc-protected primary diamine for selective deprotection.
  • Hydrochloride salt form improves stability and handling.
  • High reported purity for synthesis and analytical use.
  • Available in small lab-scale pack sizes for research use.
  • Characterized by CAS number and supported by certificate of analysis.