Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical N-Fmoc-1,5-diaminopentane HCl | 5 g | CAS 177333-17-4 | MDL MFCD00832511

Catalog No. 502345135
Encompass

N-Fmoc-1,5-diaminopentane HCl is a Amino Acid reagent (Subcategory: Building Block) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 177333-17-4
- MDL: MFCD00832511
- InChIKey: MYVDRDMFYHOZBJ-UHFFFAOYSA-N
- Molecular Weight: 360.882
- Molecular Formula: C20H25ClN2O2
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (9H-fluoren-9-yl)methyl (5-aminopentyl)carbamate hydrochloride
- SMILES: NCCCCCNC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O.Cl

Catalog No. 50-234-5135 Supplier STA Pharmaceutical Supplier No. STAAA116985GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Fmoc-1,5-diaminopentane HCl is a Amino Acid reagent (Subcategory: Building Block) sold by WuXi TIDES. Offered in 5 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 177333-17-4
- MDL: MFCD00832511
- InChIKey: MYVDRDMFYHOZBJ-UHFFFAOYSA-N
- Molecular Weight: 360.882
- Molecular Formula: C20H25ClN2O2
- Purity: ≥95%
- Container Type: 30 mL HDPE
- Pack Size: 5 g
- Net Weight: 5 g
- Gross Weight: 16.5 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (9H-fluoren-9-yl)methyl (5-aminopentyl)carbamate hydrochloride
- SMILES: NCCCCCNC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O.Cl

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.