Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical N-Fmoc-2-amino-2-(2-propenyl)-4-Pentenoic acid | 25 g | CAS 1311992-97-8 | MDL MFCD17215634

Catalog No. 502362128
Encompass

N-Fmoc-2-amino-2-(2-propenyl)-4-Pentenoic acid is a Amino Acid reagent (Subcategory: Alkenyl AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1311992-97-8
- MDL: MFCD17215634
- InChIKey: HNFDPUQNBJDSGT-UHFFFAOYSA-N
- Molecular Weight: 377.44
- Molecular Formula: C23H23NO4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-allylpent-4-enoic acid
- SMILES: C=CCC(NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)(CC=C)C(O)=O

Catalog No. 50-236-2128 Supplier STA Pharmaceutical Supplier No. STAAA1133725GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Fmoc-2-amino-2-(2-propenyl)-4-Pentenoic acid is a Amino Acid reagent (Subcategory: Alkenyl AA) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 1311992-97-8
- MDL: MFCD17215634
- InChIKey: HNFDPUQNBJDSGT-UHFFFAOYSA-N
- Molecular Weight: 377.44
- Molecular Formula: C23H23NO4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-2-allylpent-4-enoic acid
- SMILES: C=CCC(NC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)(CC=C)C(O)=O

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.