Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-2-cyclopentyl-L-glycine, 95%

Catalog No. AAH6615406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6615406 5 g
AAH6615403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6615406 Supplier Thermo Scientific Chemicals Supplier No. H6615406

missing translation for 'validMainframeMsg'

CAS: 220497-61-0 | C22H23NO4 | 365.429 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220497-61-0
Molecular Formula C22H23NO4
Molecular Weight (g/mol) 365.429
MDL Number MFCD01311751
InChI Key JGWHUIQSEKGLAQ-FQEVSTJZSA-N
Synonym fmoc-cpg-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-2-cyclopentylacetic acid, fmoc-cyclopentyl-gly-oh, s-cyclopentyl 9h-fluoren-9-ylmethoxy carbonyl amino acetic acid, fmoc-l-cpg-oh, ambotzfaa1360, fmoc-l-cyclopentyl-glycine, n-fmoc-alpha-cyclopentylglycine, fmoc-s-2-cyclopentylglycine, n-alpha-9-fluorenylmethyloxycarbonyl-l-beta-cyclopentylglycine
PubChem CID 7006544
IUPAC Name (2S)-2-cyclopentyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
SMILES C1CCC(C1)C(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Color White
Quantity 5 g
Formula Weight 379.46
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-2-cyclopentyl-L-glycine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.