Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-3-(2-pyridyl)-L-alanine, 95%, Thermo Scientific™

Catalog No. AAH66529MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66529MD 250 mg
AAH6652903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66529MD Supplier Thermo Scientific Chemicals Supplier No. H66529MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 185379-40-2
Molecular Formula C23H20N2O4
Molecular Weight (g/mol) 388.423
MDL Number MFCD00672564
InChI Key DXIVJCDZOMUITR-NRFANRHFSA-N
Synonym fmoc-l-2-pyridylalanine, fmoc-2-pal-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-2-yl propanoic acid, fmoc-2'-pyridyl-l-ala, fmoc-beta-2-pyridyl-ala-oh, s-n-fmoc-2-pyridyl alanine, fmoc-l-3-2-pyridyl-alanine, fmoc-l-3-2-pyridyl alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-pyridin-2-yl propanoic acid, fmoc-d-3-2-pyridyl alanine
PubChem CID 978320
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-2-ylpropanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=CC=N4)C(=O)O

Specifications

Color Brown
Quantity 250 mg
Formula Weight 388.42
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-3-(2-pyridyl)-L-alanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.