Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-4-benzoyl-L-phenylalanine, 95%

Catalog No. AAH6670706
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6670706 5 g
AAH66707MD 250 mg
AAH6670703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6670706 Supplier Thermo Scientific Chemicals Supplier No. H6670706
Only null left
Add to Cart

CAS: 117666-96-3 | C31H25NO5 | 491.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117666-96-3
Molecular Formula C31H25NO5
Molecular Weight (g/mol) 491.54
MDL Number MFCD00151937
InChI Key SYOBJKCXNRQOGA-NDEPHWFRSA-N
Synonym fmoc-bpa-oh, fmoc-l-4-benzoylphenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzoylphenyl propanoic acid, 2s-3-4-benzoylphenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, l-phenylalanine, 4-benzoyl-n-9h-fluoren-9-ylmethoxy carbonyl, fmoc-p-bz-phe-oh, fmoc-4-benzoyl-l-phenylalanine, benzoylphenylfluorenylmethoxycarbonylaminopropanoicacid, n-9h-fluorene-9-ylmethoxycarbonyl-4-benzoyl-l-phenylalanine
PubChem CID 14233360
IUPAC Name (2S)-3-(4-benzoylphenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Color White
Beilstein 7672022
Quantity 5 g
Formula Weight 491.54
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Fmoc-4-benzoyl-L-phenylalanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.