Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Fmoc-4-methyl-L-phenylalanine, 95%

Catalog No. AAH52073MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52073MD 250 mg
AAH5207303 1 g
AAH5207306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH52073MD Supplier Thermo Scientific Chemicals Supplier No. H52073MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 199006-54-7 | C25H23NO4 | 401.462 g/mol

N-Fmoc-4-methyl-L-phenylalanine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-4-methyl-L-phenylalanine is used as pharmaceutical intermediate.

Solubility
Soluble in Dimethylformamide(DMF).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 199006-54-7
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.462
MDL Number MFCD00270197
InChI Key UXLHLZHGQPDMJQ-QHCPKHFHSA-N
Synonym fmoc-phe 4-me-oh, fmoc-l-4-methylphe, fmoc-4-methyl-l-phenylalanine, fmoc-l-4-methylphenylalanine, fmoc-4-methy-l-phenylalanine, fmoc-l-phe 4-me-oh, fmoc-p-me-phe-oh, n-fmoc-4-methyl-l-phenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-p-tolyl propanoic acid, phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-methyl
PubChem CID 7129826
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-methylphenyl)propanoic acid
SMILES CC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 250 mg
Beilstein 8799411
Solubility Information Soluble in Dimethylformamide(DMF).
Formula Weight 401.46
Percent Purity 95%
Chemical Name or Material N-Fmoc-4-methyl-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.