Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Fmoc-4-(tert-butoxycarbonylmethoxy)-L-phenylalanine | 181951-92-8 | >=98.0% | 517.57 | C30H31NO7 | 5 G
SDP

Catalog No. 5000208286
Encompass_Preferred

N-Fmoc-4-(tert-butoxycarbonylmethoxy)-L-phenylalanine is an Fmoc-protected phenylalanine derivative used as a building block in peptide synthesis and related organic chemistry applications.

  • Purity ≥98.0% (NMR).
  • Molecular weight 517.57 g/mol.
  • Molecular formula C30H31NO7.
  • Appearance: powder.
  • Storage: powder -20°C for up to 3 years or 4°C for up to 2 years; in solvent -80°C for up to 6 months or -20°C for up to 1 month.
  • CAS number 181951-92-8.

Catalog No. 50-002-08286 Supplier Medchemexpress LLC Supplier No. HYW0880975G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Fmoc-4-(tert-butoxycarbonylmethoxy)-L-phenylalanine is an Fmoc-protected phenylalanine derivative used as a building block in peptide synthesis and related organic chemistry applications.

  • Purity ≥98.0% (NMR).
  • Molecular weight 517.57 g/mol.
  • Molecular formula C30H31NO7.
  • Appearance: powder.
  • Storage: powder -20°C for up to 3 years or 4°C for up to 2 years; in solvent -80°C for up to 6 months or -20°C for up to 1 month.
  • CAS number 181951-92-8.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.