Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Fmoc-4-(tert-butoxycarbonylmethoxy)-L-phenylalanine | 181951-92-8 | MFCD07781259 | ≥98.0% | 517.57 | C30H31NO7 | 250 MG
SDP

Catalog No. 5000208695
Encompass_Preferred

N-Fmoc-4-(tert-butoxycarbonylmethoxy)-L-phenylalanine is an Fmoc-protected derivative of L-phenylalanine used as a building block in peptide synthesis and related peptide chemistry applications. It features an N-Fmoc protecting group on the amine and a tert-butoxycarbonylmethoxy protecting group at the 4-position of the phenyl ring, supplied as a solid powder with reported high purity for synthetic use.

  • Fmoc-protected amino acid for solid-phase peptide synthesis.
  • Tert-butoxycarbonylmethoxy group at the 4-position provides orthogonal protection.
  • Molecular formula: C30H31NO7.
  • Molecular weight approximately 517.6 daltons.
  • Purity by NMR: ≥98.0%.
  • Solid powder form; store at -20°C for long-term stability.

Catalog No. 50-002-08695 Supplier Medchemexpress LLC Supplier No. 250MGHYW088097
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Fmoc-4-(tert-butoxycarbonylmethoxy)-L-phenylalanine is an Fmoc-protected derivative of L-phenylalanine used as a building block in peptide synthesis and related peptide chemistry applications. It features an N-Fmoc protecting group on the amine and a tert-butoxycarbonylmethoxy protecting group at the 4-position of the phenyl ring, supplied as a solid powder with reported high purity for synthetic use.

  • Fmoc-protected amino acid for solid-phase peptide synthesis.
  • Tert-butoxycarbonylmethoxy group at the 4-position provides orthogonal protection.
  • Molecular formula: C30H31NO7.
  • Molecular weight approximately 517.6 daltons.
  • Purity by NMR: ≥98.0%.
  • Solid powder form; store at -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.