Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Fmoc-4-(tert-butoxycarbonylmethoxy)-L-phenylalanine | 181951-92-8 | MFCD07781259 | >98.0% | 517.57 g/mol | C30H31NO7 | 1 G
SDP

Catalog No. 5000208555
Encompass_Preferred

N-Fmoc-4-(tert-butoxycarbonylmethoxy)-L-phenylalanine is an Fmoc-protected phenylalanine derivative used as a protected amino acid building block for peptide synthesis and chemical research. It features a tert-butoxycarbonyl-protected methoxy substituent at the 4-position of the aromatic ring and is supplied as a solid with formula C30H31NO7 and molecular weight 517.57 g/mol. Typical purity is ≥98.0% and recommended storage is powder: -20°C (up to 3 years) or 4°C (up to 2 years); in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

  • Fmoc-protected amino acid suitable for solid-phase peptide synthesis.
  • Boc-protected methoxy group at the 4-position of the aromatic ring.
  • Chemical formula C30H31NO7 and molecular weight 517.57 g/mol.
  • Purity ≥98.0% by 1H NMR.
  • Supplied as a solid; follow recommended powder and solvent storage conditions.
  • Intended for research use only.

Catalog No. 50-002-08555 Supplier Medchemexpress LLC Supplier No. HYW0880971G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Fmoc-4-(tert-butoxycarbonylmethoxy)-L-phenylalanine is an Fmoc-protected phenylalanine derivative used as a protected amino acid building block for peptide synthesis and chemical research. It features a tert-butoxycarbonyl-protected methoxy substituent at the 4-position of the aromatic ring and is supplied as a solid with formula C30H31NO7 and molecular weight 517.57 g/mol. Typical purity is ≥98.0% and recommended storage is powder: -20°C (up to 3 years) or 4°C (up to 2 years); in solvent: -80°C (up to 6 months) or -20°C (up to 1 month).

  • Fmoc-protected amino acid suitable for solid-phase peptide synthesis.
  • Boc-protected methoxy group at the 4-position of the aromatic ring.
  • Chemical formula C30H31NO7 and molecular weight 517.57 g/mol.
  • Purity ≥98.0% by 1H NMR.
  • Supplied as a solid; follow recommended powder and solvent storage conditions.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.