Learn More
Medchemexpress LLC N-Fmoc-8-aminooctanoic acid | 126631-93-4 | 98.95% | 381.46 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW009030
Encompass_Preferred
N-Fmoc-8-aminooctanoic acid is a PROTAC linker used in the synthesis of PROTACs. It is an alkane chain featuring terminal Fmoc-protected amine and carboxylic acid groups. The Fmoc group can be deprotected under basic conditions to yield a free amine, which is suitable for further conjugations. The terminal carboxylic acid can form a stable amide bond by reacting with primary amine groups in the presence of activators like EDC or HATU. This product is for research use only.
  • Can be used as a PROTAC linker.
  • Contains an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups.
  • Fmoc group can be deprotected for further conjugations.
  • Terminal carboxylic acid can form stable amide bonds with primary amine groups.
Catalog No. 50-004-54100