Learn More
Medchemexpress LLC N-Fmoc-8-aminooctanoic acid | 126631-93-4 | 99.0% | 381.46 | 5 G
SDP
Catalog No. 5000453847
Encompass_Preferred
N-Fmoc-8-aminooctanoic acid can be used as a PROTAC linker in the synthesis of PROTACs. It is an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups. The Fmoc group can be deprotected under basic conditions to obtain a free amine for further conjugations. The terminal carboxylic acid can react with primary amine groups in the presence of activators (e.g., EDC or HATU) to form a stable amide bond.
  • Can be used as a PROTAC linker
  • Alkane chain with terminal Fmoc-protected amine and carboxylic acid groups
  • Fmoc group can be deprotected under basic conditions
  • Terminal carboxylic acid can react with primary amine groups to form a stable amide bond
Catalog No. 50-004-53847 Supplier Medchemexpress LLC Supplier No. HYW0090305G
N-Fmoc-8-aminooctanoic acid can be used as a PROTAC linker in the synthesis of PROTACs. It is an alkane chain with terminal Fmoc-protected amine and carboxylic acid groups. The Fmoc group can be deprotected under basic conditions to obtain a free amine for further conjugations. The terminal carboxylic acid can react with primary amine groups in the presence of activators (e.g., EDC or HATU) to form a stable amide bond.
  • Can be used as a PROTAC linker
  • Alkane chain with terminal Fmoc-protected amine and carboxylic acid groups
  • Fmoc group can be deprotected under basic conditions
  • Terminal carboxylic acid can react with primary amine groups to form a stable amide bond